2-Methyl-5-nitrobenzotrifluorid
Lieferant: Apollo Scientific
|
Gefahr
|
PC0422-25G
PC0422-5G
PC0422-100G
APOSPC0422-25GEA
58
EUR
APOSPC0422-25G
APOSPC0422-5G
APOSPC0422-100G
2-Methyl-5-nitrobenzotrifluorid
2-Methyl-5-nitrobenzotrifluorid
Formel:
C₈H₆F₃NO₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD01631684 CAS-Nummer: 89976-12-5 UN: 3439 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...