5-Fluor-2-(trifluormethyl)benzamid 97%

Lieferant: Apollo Scientific

PC0425-5G PC0425-1G
APOSPC0425-5GEA 104 EUR
APOSPC0425-5G APOSPC0425-1G
5-Fluor-2-(trifluormethyl)benzamid 97%
5-Fluor-2-(trifluormethyl)benzamid
Formel: C₈H₅F₄NO
Lagertemperatur: Raumtemperatur
CAS-Nummer: 654-95-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR