5-Fluor-2-(trifluormethyl)benzamid 97%
Lieferant: Apollo Scientific
PC0425-5G
PC0425-1G
APOSPC0425-5GEA
104
EUR
APOSPC0425-5G
APOSPC0425-1G
5-Fluor-2-(trifluormethyl)benzamid 97%
5-Fluor-2-(trifluormethyl)benzamid
Formel:
C₈H₅F₄NO Lagertemperatur: Raumtemperatur |
CAS-Nummer:
654-95-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...