N-Fmoc-2-fluoro-L-phenylalanine

Lieferant: Apollo Scientific
Achtung

Synonyme: (S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(2-fluorophenyl)propanoic acid, Fmoc-Phe(2-F)-OH

PC0426-25G
APOSPC0426-25GEA 201 EUR
APOSPC0426-25G
N-Fmoc-2-fluoro-L-phenylalanine
N-Fmoc-2-fluoro-L-phenylalanine
Formel: C₂₄H₂₀FNO₄
Molecular Weight: 405.425 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00672552
CAS-Nummer: 205526-26-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR