3-Methyl-5-(trifluoromethyl)benzamide

Lieferant: Apollo Scientific

PC0442-10G PC0442-5G
APOSPC0442-10GEA 0 EUR
APOSPC0442-10G APOSPC0442-5G
3-Methyl-5-(trifluoromethyl)benzamide
3-Methyl-5-(trifluoromethyl)benzamide
Formel: C₉H₈F₃NO
Lagertemperatur: Raumtemperatur
MDL: MFCD01631497
CAS-Nummer: 261951-98-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR