2-Methyl-3-(trifluoromethyl)benzamide 97%

Lieferant: Apollo Scientific

PC0445-1G
APOSPC0445-1GEA 116 EUR
APOSPC0445-1G
2-Methyl-3-(trifluoromethyl)benzamide 97%
2-Methyl-3-(trifluoromethyl)benzamide
Formel: C₉H₈F₃NO
Lagertemperatur: Raumtemperatur
MDL: MFCD01631495
CAS-Nummer: 251651-26-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR