4-Methyl-2-(trifluormethyl)benzoesäure

Lieferant: Apollo Scientific
Achtung

PC0449-5G PC0449-10G PC0449-1G PC0449-25G
APOSPC0449-5GEA 101 EUR
APOSPC0449-5G APOSPC0449-10G APOSPC0449-1G APOSPC0449-25G
4-Methyl-2-(trifluormethyl)benzoesäure
4-Methyl-2-(trifluoromethyl)benzoic acid
Formel: C₉H₇F₃O₂
Molecular Weight: 204.15 g/mol
Schmelzpunkt: 135…141 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01631591
CAS-Nummer: 120985-64-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR