Chlor(di-1-pyrrolidinyl)methyliumhexafluorophosphat

Lieferant: Apollo Scientific

Synonyme: 1-(Chloro(pyrrolidin-1-yl)methylene)pyrrolidin-1-ium hexafluorophosphate(V), Chloro-N,N,N',N'-bis(tetramethylene)formamidinium Hexafluorophosphate, PyClU

PC0451-25G
APOSPC0451-25GEA 88.6 EUR
APOSPC0451-25G
Chlor(di-1-pyrrolidinyl)methyliumhexafluorophosphat
1-(Chloro-1-pyrrolidinylmethylene)pyrrolidinium hexafluorophosphate
Formel: C₉H₁₆ClF₆N₂P
Molecular Weight: 332,66 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00191333
CAS-Nummer: 135540-11-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR