4-Methylthio-3-nitrobenzotrifluorid
Lieferant: Apollo Scientific
PC0463-5G
APOSPC0463-5GEA
154
EUR
APOSPC0463-5G
4-Methylthio-3-nitrobenzotrifluorid
4-Methylthio-3-nitrobenzotrifluorid
Formel:
C₈H₆F₃NO₂S Schmelzpunkt: 94…96 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00052206 CAS-Nummer: 71236-96-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...