2-Methyl-5-(trifluoromethyl)benzyl alcohol

Lieferant: Apollo Scientific
Achtung

PC0474-500MG
APOSPC0474-500MGEA 21.9 EUR
APOSPC0474-500MG
2-Methyl-5-(trifluoromethyl)benzyl alcohol
2-Methyl-5-(trifluoromethyl)benzyl alcohol
Formel: C₉H₉F₃O
Lagertemperatur: Raumtemperatur
MDL: MFCD01631604
CAS-Nummer: 261952-13-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR