3-Methyl-5-(trifluoromethyl)benzyl alcohol 98%

Lieferant: Apollo Scientific

PC0476-1G
APOSPC0476-1GEA 0 EUR
APOSPC0476-1G
3-Methyl-5-(trifluoromethyl)benzyl alcohol 98%
3-Methyl-5-(trifluoromethyl)benzyl alcohol
Formel: C₉H₉F₃O
Lagertemperatur: Raumtemperatur
MDL: MFCD01631605
CAS-Nummer: 116070-38-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR