4-Methyl-3-(trifluoromethyl)benzyl alcohol

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-(Hydroxymethyl)-2-methylbenzotrifluoride, [4-Methyl-3-(trifluoromethyl)phenyl]methanol

PC0478-250MG
APOSPC0478-250MGEA 29 EUR
APOSPC0478-250MG
4-Methyl-3-(trifluoromethyl)benzyl alcohol
4-Methyl-3-(trifluoromethyl)benzyl alcohol
Formel: C₉H₉F₃O
Molecular Weight: 190,162 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01631607
CAS-Nummer: 261952-15-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR