2-Methyl-3-(trifluormethyl)benzylbromid 98%

Lieferant: Apollo Scientific
Gefahr

Synonyme: 1-(Bromomethyl)-2-methyl-3-(trifluoromethyl)benzene

PC0479-1G PC0479-5G PC0479-10G PC0479-25G PC0479-100G
APOSPC0479-1GEA 0 EUR
APOSPC0479-1G APOSPC0479-5G APOSPC0479-10G APOSPC0479-25G APOSPC0479-100G
2-Methyl-3-(trifluormethyl)benzylbromid 98%
2-Methyl-3-(trifluormethyl)benzylbromid
Formel: C₉H₈BrF₃
Molecular Weight: 253.06 g/mol
Siedepunkt: 72 °C/1mmHg
Lagertemperatur: Raumtemperatur
MDL: MFCD01631608
CAS-Nummer: 261952-16-3
UN: 3265
ADR: 8 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR