2-Methyl-5-(trifluormethyl)benzylbromid 98%

Lieferant: Apollo Scientific
Gefahr

PC0480-1G PC0480-5G PC0480-10G
APOSPC0480-1GEA 34.8 EUR
APOSPC0480-1G APOSPC0480-5G APOSPC0480-10G
2-Methyl-5-(trifluormethyl)benzylbromid 98%
2-Methyl-5-(trifluormethyl)benzylbromid
Formel: C₉H₈BrF₃
Lagertemperatur: Raumtemperatur
MDL: MFCD01631609
CAS-Nummer: 261952-17-4
UN: 3265
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR