3-Methyl-5-(trifluormethyl)benzylbromid

Lieferant: Apollo Scientific
Gefahr

PC0483-1G PC0483-5G
APOSPC0483-1GEA 0 EUR
APOSPC0483-1G APOSPC0483-5G
3-Methyl-5-(trifluormethyl)benzylbromid
3-Methyl-5-(trifluormethyl)benzylbromid
Formel: C₉H₈BrF₃
Lagertemperatur: Raumtemperatur
MDL: MFCD01631610
CAS-Nummer: 116070-36-1
UN: 3265
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR