2,3,4,5-Tetrafluorbenzamid 97%

Lieferant: Apollo Scientific

Synonyme: 2,3,4,5-Tetrafluorbenzoesäureamid

PC0496-1G
APOSPC0496-1GEA 21.9 EUR
APOSPC0496-1G
2,3,4,5-Tetrafluorbenzamid 97%
2,3,4,5-Tetrafluorbenzamid
Formel: C₇H₃F₄NO
Schmelzpunkt: 132…134 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01631457
CAS-Nummer: 16582-94-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR