Dimethylphenacylsulfoniumtetrafluoroborat 98%
Lieferant: Apollo Scientific
PC0503-1G
APOSPC0503-1GEA
154
EUR
APOSPC0503-1G
Dimethylphenacylsulfoniumtetrafluoroborat 98%
Dimethylphenacylsulfoniumtetrafluoroborat
Formel:
C₁₀H₁₃BF₄OS Schmelzpunkt: 170 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00191352 CAS-Nummer: 24806-57-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...