Fmoc-3,4,5-trifluoro-D-phenylalanine
Lieferant: Apollo Scientific
Synonyme:
(R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(3,4,5-trifluorophenyl)propanoic acid, Fmoc-D-Phe(3,4,5-DiF)-OH
PC0504-1G
APOSPC0504-1GEA
214
EUR
APOSPC0504-1G
Fmoc-3,4,5-trifluoro-D-phenylalanine
Fmoc-3,4,5-trifluoro-D-phenylalanine
Formel:
C₂₄H₁₈F₃NO₄ Molecular Weight: 441.41 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00797584 CAS-Nummer: 205526-31-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...