Perfluoro-4-methylbenzoic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Carboxy-2,3,5,6-tetrafluorobenzotrifluoride, 2,3,5,6-Tetrafluoro-4-(trifluoromethyl)benzoic acid

PC0506-5G
APOSPC0506-5GEA 154 EUR
APOSPC0506-5G
Perfluoro-4-methylbenzoic acid
Perfluoro-4-methylbenzoic acid
Formel: C₈HF₇O₂
Molecular Weight: 262,081 g/mol
Schmelzpunkt: 109…110 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01631500
CAS-Nummer: 5216-22-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR