5-(Trifluoromethyl)dibenzothiophenium trifluoromethanesulphonate 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-(Trifluoromethyl)-5H-dibenzo[b,d]thiophen-5-ium trifluoromethanesulphonate

PC0512-1G PC0512-5G
APOSPC0512-1GEA 68.3 EUR
APOSPC0512-1G APOSPC0512-5G
5-(Trifluoromethyl)dibenzothiophenium trifluoromethanesulphonate 97%
5-(Trifluoromethyl)dibenzothiophenium trifluoromethanesulphonate

Trifluoromethylating agent. Technical bulletin available.

Formel: C₁₄H₈F₆O₃S₂
Schmelzpunkt: 150 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00236132
CAS-Nummer: 129946-88-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR