2,3,4-Trifluoro-6-(trifluoromethyl)benzoic acid

Lieferant: Apollo Scientific
Achtung

PC0534-1G PC0534-250MG
APOSPC0534-1GEA 0 EUR
APOSPC0534-1G APOSPC0534-250MG
2,3,4-Trifluoro-6-(trifluoromethyl)benzoic acid
2,3,4-Trifluoro-6-(trifluoromethyl)benzoic acid
Formel: C₈H₂F₆O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD01631432
CAS-Nummer: 157337-84-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR