(3-Chlor-5-fluorphenyl)essigsäure

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-(3-Chloro-5-fluorophenyl)acetic acid

PC0538-1G PC0538-25G PC0538-5G PC0538-100G PC0538-500G
APOSPC0538-1GEA 21.9 EUR
APOSPC0538-1G APOSPC0538-25G APOSPC0538-5G APOSPC0538-100G APOSPC0538-500G
(3-Chlor-5-fluorphenyl)essigsäure
(3-Chlor-5-fluorphenyl)essigsäure
Formel: (C₆H₃ClF)CH₂COOH
Molecular Weight: 188.59 g/mol
Schmelzpunkt: 82…85 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03788475
CAS-Nummer: 202001-00-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR