3,5-Bis(trifluormethyl)phenylboronsäure 98%
Lieferant: Apollo Scientific
|
Achtung
|
PC0540-25G
PC0540-5G
PC0540-100G
APOSPC0540-25GEA
0
EUR
APOSPC0540-25G
APOSPC0540-5G
APOSPC0540-100G
3,5-Bis(trifluormethyl)phenylboronsäure 98%
3,5-Bis(trifluormethyl)phenylboronsäure
Formel:
C₈H₅BF₆O₂ Schmelzpunkt: 217…221 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00051850 CAS-Nummer: 73852-19-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...