Ethyl 1-methyl-5-(trifluoromethyl)-1H-pyrazole-4-carboxylate 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-(Ethoxycarbonyl)-1-methyl-5-(trifluoromethyl)-1H-pyrazole
PC0541-1G
APOSPC0541-1GEA
78.3
EUR
APOSPC0541-1G
Ethyl 1-methyl-5-(trifluoromethyl)-1H-pyrazole-4-carboxylate 97%
Ethyl 1-methyl-5-(trifluoromethyl)-1H-pyrazole-4-carboxylate
Formel:
C₈H₉F₃N₂O₂ Schmelzpunkt: 58…61 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00277430 CAS-Nummer: 231285-86-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...