4-(Trifluoromethyl)cinnamamide 98%
Lieferant: Apollo Scientific
|
Achtung
|
PC0552-1G
PC0552-5G
APOSPC0552-1GEA
0
EUR
APOSPC0552-1G
APOSPC0552-5G
4-(Trifluoromethyl)cinnamamide 98%
4-(Trifluoromethyl)cinnamamide
Formel:
C₁₀H₈F₃NO Lagertemperatur: Kühlschrank |
MDL:
MFCD03092988 CAS-Nummer: 115093-99-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...