4-(Trifluoromethyl)cinnamamide 98%

Lieferant: Apollo Scientific
Achtung

PC0552-1G PC0552-5G
APOSPC0552-1GEA 0 EUR
APOSPC0552-1G APOSPC0552-5G
4-(Trifluoromethyl)cinnamamide 98%
4-(Trifluoromethyl)cinnamamide
Formel: C₁₀H₈F₃NO
Lagertemperatur: Kühlschrank
MDL: MFCD03092988
CAS-Nummer: 115093-99-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR