4-(4-Fluoro-3-methylphenyl)-1,3-thiazol-2-amine 98%
Lieferant: Apollo Scientific
|
Achtung
|
PC0569-500MG
APOSPC0569-500MGEA
21.9
EUR
APOSPC0569-500MG
4-(4-Fluoro-3-methylphenyl)-1,3-thiazol-2-amine 98%
4-(4-Fluoro-3-methylphenyl)-1,3-thiazol-2-amine
Formel:
C₁₀H₉FN₂S Schmelzpunkt: 101…103 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00042158 CAS-Nummer: 3830-48-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...