Heptafluoroisopropyl acrylate
Lieferant: Apollo Scientific
|
Achtung
|
PC0572-1G
APOSPC0572-1GEA
84.6
EUR
APOSPC0572-1G
Heptafluoroisopropyl acrylate
Heptafluoroisopropyl acrylate
Formel:
C₆H₃F₇O₂ Siedepunkt: 86…87 °C Dichte: 1,4 g/cm³ (25 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00274356 CAS-Nummer: 13057-08-4 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...