3'-Fluor-4'-hydroxyacetophenon

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-(3-Fluoro-4-hydroxyphenyl)-1-ethanone

PC0584-1G
APOSPC0584-1GEA 21.9 EUR
APOSPC0584-1G
3'-Fluor-4'-hydroxyacetophenon
3'-Fluor-4'-hydroxyacetophenon
Formel: C₈H₇FO₂
Molecular Weight: 154.14 g/mol
Schmelzpunkt: 127…129 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00215840
CAS-Nummer: 403-14-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR