2-Hydroxy-5-(trifluoromethoxy)phenacyl bromide

Lieferant: Apollo Scientific

PC0594-1G
APOSPC0594-1GEA 408 EUR
APOSPC0594-1G
2-Hydroxy-5-(trifluoromethoxy)phenacyl bromide
2-Hydroxy-5-(trifluoromethoxy)phenacyl bromide
Formel: C₉H₆BrF₃O₃
Schmelzpunkt: 43…45 °C
Lagertemperatur: Kühlschrank
MDL: MFCD07366522
CAS-Nummer: 496052-53-0
UN: 1760
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR