(1S)-1-[3-(Trifluoromethyl)phenyl]ethylamine

Lieferant: Apollo Scientific
Achtung

Synonyme: (S)-1-(3-(Trifluoromethyl)phenyl)ethanamine

PC0600-50MG
APOSPC0600-50MGEA 36.4 EUR
APOSPC0600-50MG
(1S)-1-[3-(Trifluoromethyl)phenyl]ethylamine
(1S)-1-[3-(Trifluoromethyl)phenyl]ethylamine
Formel: C₉H₁₀F₃N
Molecular Weight: 189,178 g/mol
MDL: MFCD03093006
CAS-Nummer: 127852-21-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR