(1S)-1-[4-(Trifluoromethyl)phenyl]ethylamine 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(S)-α-Methyl-4-(trifluoromethyl)benzylamine
PC0601-1G
PC0601-250MG
APOSPC0601-1GEA
212
EUR
APOSPC0601-1G
APOSPC0601-250MG
(1S)-1-[4-(Trifluoromethyl)phenyl]ethylamine 98%
(1S)-1-[4-(Trifluoromethyl)phenyl]ethylamine
Formel:
C₉H₁₀F₃N Molecular Weight: 189.178 g/mol Siedepunkt: 76 °C (20 mm Hg) Dichte: 1.03 g/cm³ (25 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD03093009 CAS-Nummer: 84499-73-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...