(1S)-1-(2-Fluorophenyl)ethan-1-ol
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(S)-2-Fluoro-α-methylbenzyl alcohol, (S)-1-(2-Fluorophenyl)ethanol
PC0605-1G
APOSPC0605-1GEA
60.9
EUR
APOSPC0605-1G
(1S)-1-(2-Fluorophenyl)ethan-1-ol
(1S)-1-(2-Fluorophenyl)ethan-1-ol
Formel:
C₈H₉FO Molecular Weight: 140.155 g/mol Siedepunkt: 110 °C ( 135 mm Hg) Dichte: 1.1 g/cm³ (25 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD03092992 CAS-Nummer: 171032-87-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...