(R)-1-(3-Fluorophenyl)ethanol 95%

Lieferant: Apollo Scientific

PC0606-5G
APOSPC0606-5GEA 209 EUR
APOSPC0606-5G
(R)-1-(3-Fluorophenyl)ethanol 95%
(R)-1-(3-Fluorophenyl)ethanol
Formel: C₈H₉FO
Molecular Weight: 140.155 g/mol
MDL: MFCD03092995
CAS-Nummer: 126534-33-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR