(S)-(+)-1,1'-Binaphthyl-2,2'-diylbis(trifluoromethanesulphonate)

Lieferant: Apollo Scientific
Gefahr

Synonyme: (S)-(+)-1,1'-Bi-2-naphthol bis(trifluoromethanesulphonate), (S)-[1,1-Binaphthalene]-2,2-diyl bis(trifluoromethanesulphonate), (S)-(+)-1,1'-Bi-2-naphthol Bis(trifluoromethanesulphonate)

PC0615-25G
APOSPC0615-25GEA 195 EUR
APOSPC0615-25G
(S)-(+)-1,1'-Binaphthyl-2,2'-diylbis(trifluoromethanesulphonate)
(S)-(+)-1,1'-Binaphthyl-2,2'-diylbis(trifluoromethanesulphonate)
Formel: C₂₂H₁₂F₆O₆S₂
Schmelzpunkt: 83…85 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00274615
CAS-Nummer: 128544-05-8
UN: 3261
ADR: 8,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR