(3S)-4,4,4-Trifluorobutane-1,3-diol 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(2S)-2,4-Dihydroxy-1,1,1-trifluorobutane, (S)-4,4,4-Trifluorobutane-1,3-diol, S-TFBDOL
PC0617-250MG
APOSPC0617-250MGEA
655
EUR
APOSPC0617-250MG
(3S)-4,4,4-Trifluorobutane-1,3-diol 97%
(3S)-4,4,4-Trifluorobutane-1,3-diol
Formel:
C₄H₇F₃O₂ Molecular Weight: 144,092 g/mol Siedepunkt: 75…76 °C (5 mm Hg) Lagertemperatur: Raumtemperatur |
MDL:
MFCD03093015 CAS-Nummer: 135154-88-0 UN: 1993 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...