3,3,3-Trifluoro-D-lactic acid 97%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
(2R)-2-Hydroxy-3,3,3-trifluoropropanoic acid, (R)-3,3,3-Trifluoro-2-hydroxypropanoic acid
PC0618-1G
PC0618-250MG
APOSPC0618-1GEA
630
EUR
APOSPC0618-1G
APOSPC0618-250MG
3,3,3-Trifluoro-D-lactic acid 97%
3,3,3-Trifluoro-D-lactic acid
Formel:
C₃H₃F₃O₃ Molecular Weight: 144.049 g/mol Schmelzpunkt: 66…71 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03093016 CAS-Nummer: 121250-04-2 UN: 3261 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...