5-Cyan-2-fluorphenylboronsäure 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
5-Cyano-2-fluorobenzeneboronic acid
PC062-1G
APOSPC062-1GEA
21.9
EUR
APOSPC062-1G
5-Cyan-2-fluorphenylboronsäure 98%
5-Cyan-2-fluorphenylboronsäure
Formel:
C₇H₅BFNO₂ Molecular Weight: 164,9300 g/mol Schmelzpunkt: 300 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD03095147 CAS-Nummer: 468718-30-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...