4-Fluor-D-phenylalanin

Lieferant: Apollo Scientific

Synonyme: (R)-2-Amino-3-(4-fluorophenyl)propanoic acid, H-D-Phe(4-F)-OH

PC0660-5G
APOSPC0660-5GEA 66.7 EUR
APOSPC0660-5G
4-Fluor-D-phenylalanin
4-Fluor-D-phenylalanin
Formel: C₉H₁₀FNO₂
Molecular Weight: 183.1800 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00063063
CAS-Nummer: 18125-46-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR