2-(Trifluormethyl)phenylboronsäure 98%
Lieferant: Apollo Scientific
|
Achtung
|
PC0677-100G
PC0677-25G
PC0677-5G
APOSPC0677-100GEA
130
EUR
APOSPC0677-100G
APOSPC0677-25G
APOSPC0677-5G
2-(Trifluormethyl)phenylboronsäure 98%
2-(Trifluormethyl)phenylboronsäure
Formel:
C₇H₆BF₃O₂ Schmelzpunkt: 108…114 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00236059 CAS-Nummer: 1423-27-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...