Ethyl-4,4-difluorocyclohexanecarboxylate

Lieferant: Apollo Scientific
Achtung

Synonyme: Ethyl 4,4-difluorocyclohexanecarboxylate

PC0689-1G PC0689-5G
APOSPC0689-1GEA 21.9 EUR
APOSPC0689-1G APOSPC0689-5G
Ethyl-4,4-difluorocyclohexanecarboxylate
Ethyl-4,4-difluorocyclohexanecarboxylate
Formel: C₉H₁₄F₂O₂
Siedepunkt: 84…86 °C/8mmHg
Lagertemperatur: Raumtemperatur
MDL: MFCD03788494
CAS-Nummer: 178312-47-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR