2-Ethoxy-5-fluoro-4-pyrimidinone

Lieferant: Apollo Scientific
Achtung

Synonyme: Fluorouracil Related Compound F, 2-Ethoxy-5-fluoropyrimidin-4(3H)-one, 5-Fluoro-2-ethoxy-4-pyrimidinone

PC0704-25G
APOSPC0704-25GEA 65.4 EUR
APOSPC0704-25G
2-Ethoxy-5-fluoro-4-pyrimidinone
2-Ethoxy-5-fluoro-4-pyrimidinone
Formel: C₆H₇FN₂O₂
Molecular Weight: 158.13 g/mol
Schmelzpunkt: 180…184 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03265436
CAS-Nummer: 56177-80-1
EINECS: 260-029-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR