4-Fluorchalkon
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-Fluorbenzylidenacetophenon
PC0717-1G
APOSPC0717-1GEA
21.9
EUR
APOSPC0717-1G
4-Fluorchalkon
4-Fluorchalkon
Formel:
C₁₅H₁₁FO Molecular Weight: 226,25 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00443467 CAS-Nummer: 1608-51-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...