Pyridiniumfluorochromat 98%

Lieferant: Apollo Scientific
Gefahr

PC0727-25G PC0727-5G
APOSPC0727-25GEA 154 EUR
APOSPC0727-25G APOSPC0727-5G
Pyridiniumfluorochromat 98%
Pyridinium fluorochromate
Formel: C₅H₆CrFNO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD00151617
CAS-Nummer: 83042-08-4
UN: 1479
ADR: 5.1,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR