(R,R)-Ethylenebis(4,5,6,7-tetrahydroinden-1-yl)difluorotitanium(IV)

Lieferant: Apollo Scientific
Gefahr

PC0758-100MG
APOSPC0758-100MGEA 627 EUR
APOSPC0758-100MG
(R,R)-Ethylenebis(4,5,6,7-tetrahydroinden-1-yl)difluorotitanium(IV)
(R,R)-Ethylenebis(4,5,6,7-tetrahydroinden-1-yl)difluorotitanium(IV)

Precatalyst for the highly enantioselective imine hydrosilylation

Formel: C₂₀H₂₄F₂Ti
Molecular Weight: 350,268 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01863621
CAS-Nummer: 214361-86-1
UN: 1759

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR