Aflunox™ 14013

Lieferant: Apollo Scientific

PC0767-100G PC0767-25G PC0767-5G
APOSPC0767-100GEA 669 EUR
APOSPC0767-100G APOSPC0767-25G APOSPC0767-5G
Aflunox™ 14013
Aflunox™

Aflunox oils are clear colourless oils suitable for use as lubricants and are inert to oxidising agents such as fluorine, chlorine and oxygen at high temperatures.


  • F[CF(CF3)CF2O]xC2F5 x = 65
  • viscosity 1400 cSt@20C


Aflunox is a registered trademark of Japan Mektron

Schmelzpunkt: –30 °C
Dichte: 1.9 g/cm3 @ 25°C
MDL: MFCD00166448
CAS-Nummer: 52700-35-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR