Aflunox™ 14013
Lieferant: Apollo Scientific
PC0767-100G
PC0767-25G
PC0767-5G
APOSPC0767-100GEA
669
EUR
APOSPC0767-100G
APOSPC0767-25G
APOSPC0767-5G
Aflunox™ 14013
Aflunox™
Aflunox oils are clear colourless oils suitable for use as lubricants and are inert to oxidising agents such as fluorine, chlorine and oxygen at high temperatures.
- F[CF(CF3)CF2O]xC2F5 x = 65
- viscosity 1400 cSt@20C
Aflunox is a registered trademark of Japan Mektron
Schmelzpunkt:
–30 °C Dichte: 1.9 g/cm3 @ 25°C |
MDL:
MFCD00166448 CAS-Nummer: 52700-35-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...