N-Trifluoroacetyl-L-cysteine methyl ester 95%

Lieferant: Apollo Scientific
Achtung

PC0770-1G PC0770-250MG
APOSPC0770-1GEA 1050 EUR
APOSPC0770-1G APOSPC0770-250MG
N-Trifluoroacetyl-L-cysteine methyl ester 95%
N-Trifluoroacetyl-L-cysteine methyl ester
Formel: C₆H₈F₃NO₃S
Molecular Weight: 231.193 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00055581
CAS-Nummer: 1577-62-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR