1-(4-Fluorophenyl)-3-(2-hydroxyphenyl)prop-2-en-1-one
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4'-Fluoro-2-hydroxychalcone
PC0778-1G
APOSPC0778-1GEA
84.1
EUR
APOSPC0778-1G
1-(4-Fluorophenyl)-3-(2-hydroxyphenyl)prop-2-en-1-one
1-(4-Fluorophenyl)-3-(2-hydroxyphenyl)prop-2-en-1-one
Formel:
C₁₅H₁₁FO₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD03788512 CAS-Nummer: 76626-08-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...