4-(Pentafluorthio)anilin 98+%
Lieferant: Apollo Scientific
|
Gefahr
|
PC0780-1G
PC0780-5G
PC0780-25G
APOSPC0780-1GEA
0
EUR
APOSPC0780-1G
APOSPC0780-5G
APOSPC0780-25G
4-(Pentafluorthio)anilin 98+%
4-(Pentafluorthio)anilin
Formel:
C₆H₆F₅NS Schmelzpunkt: 64…67 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD01076203 CAS-Nummer: 2993-24-0 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...